ENA448968680 Enamine

2-benzoyl-4-methoxyaniline

  • MDL number MFCD20170481
  • Cas Number : 17549-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212837

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212837
Availability details for
Quantity
  • Availability
  • Add to Cart