ENA448968688 Enamine

2-methyl-2-(methylamino)propanamide

  • MDL number MFCD12151128
  • Cas Number : 23232-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79-81
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212942

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212942
Availability details for
Quantity
  • Availability
  • Add to Cart