ENA448968690 Enamine

4-ethenylbenzene-1-sulfonyl chloride

  • MDL number MFCD00080678
  • Cas Number : 2633-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212944

Safety Information

signalword Danger
pcodes P260,P261,P264,P272,P280,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P333+P313,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-212944
pictograms GHS05,GHS07
hstatements H314,H317
Availability details for
Quantity
  • Availability
  • Add to Cart