ENA448968697 Enamine

2-chloro-4-formylbenzene-1-sulfonamide

  • MDL number MFCD28384775
  • Cas Number : 1289063-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212953

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212953
Availability details for
Quantity
  • Availability
  • Add to Cart