ENA448968700 Enamine

3-bromo-1-methyl-1,2-dihydropyridin-2-one

  • MDL number MFCD11226865
  • Cas Number : 81971-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105978

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105978
Availability details for
Quantity
  • Availability
  • Add to Cart