ENA448968711 Enamine

methyl 2-cyano-2-(3,4-dimethoxyphenyl)acetate

  • MDL number MFCD19322173
  • Cas Number : 98466-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212995

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212995
Availability details for
Quantity
  • Availability
  • Add to Cart