ENA448968719 Enamine

3-bromo-5H,6H,7H-cyclopenta[b]pyridin-1-ium-1-olate

  • MDL number MFCD26383578
  • Cas Number : 158331-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-197534

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197534
Availability details for
Quantity
  • Availability
  • Add to Cart