ENA448968720 Enamine

N,2,2-trimethyl-3-phenoxycyclobutan-1-amine

  • MDL number MFCD21757526
  • Cas Number : 1508545-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213020

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213020
Availability details for
Quantity
  • Availability
  • Add to Cart