ENA448968724 Enamine

methyl 2-amino-3-(dimethylamino)propanoate dihydrochloride

  • MDL number MFCD28505918
  • Cas Number : 403712-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213045

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213045
Availability details for
Quantity
  • Availability
  • Add to Cart