ENA448968727 Enamine

ethyl 2-[(2-amino-4-chlorophenyl)sulfanyl]propanoate

  • MDL number MFCD16089618
  • Cas Number : 1250689-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213050

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213050
Availability details for
Quantity
  • Availability
  • Add to Cart