ENA448968731 Enamine

tert-butyldimethylsilyl 2-[(tert-butyldimethylsilyl)oxy]acetate

  • MDL number MFCD20265708
  • Cas Number : 67226-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-197604

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197604
Availability details for
Quantity
  • Availability
  • Add to Cart