ENA448968738 Enamine

5-bromo-1,2-dihydroquinolin-2-one

  • MDL number MFCD11108678
  • Cas Number : 99465-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-197731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197731
Availability details for
Quantity
  • Availability
  • Add to Cart