ENA448968751 Enamine

4-bromothiophene-2-sulfonamide

  • MDL number MFCD21893155
  • Cas Number : 214342-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213121

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213121
Availability details for
Quantity
  • Availability
  • Add to Cart