ENA448968898 Enamine

3-(bromomethyl)-1H-pyrazole hydrobromide

  • MDL number MFCD28505925
  • Cas Number : 1803590-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213142

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213142
Availability details for
Quantity
  • Availability
  • Add to Cart