ENA448968899 Enamine

(2S)-2-[(2-phenylpropyl)amino]propan-1-ol hydrochloride

  • MDL number MFCD28505927
  • Cas Number : 1807914-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213153

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213153
Availability details for
Quantity
  • Availability
  • Add to Cart