ENA448968909 Enamine

tert-butyl 5-(aminomethyl)-1,2,3,4-tetrahydroquinoline-1-carboxylate

  • MDL number MFCD28505941
  • Cas Number : 1803595-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213303

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213303
Availability details for
Quantity
  • Availability
  • Add to Cart