ENA448968931 Enamine

3-(chloromethyl)-4-methoxypyridine hydrochloride

  • MDL number MFCD28505976
  • Cas Number : 1803604-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213700

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213700
Availability details for
Quantity
  • Availability
  • Add to Cart