ENA448968936 Enamine

(1r,4r)-4-phenylcyclohexyl methanesulfonate

  • MDL number MFCD28505981
  • Cas Number : 110458-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213720

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213720
Availability details for
Quantity
  • Availability
  • Add to Cart