ENA448968937 Enamine

2-[3-(trifluoromethoxy)phenyl]morpholine

  • MDL number MFCD28505985
  • Cas Number : 1803604-31-0
  • Molecular Weight : 247.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-213732

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213732
Availability details for
Quantity
  • Availability
  • Add to Cart