ENA448968946 Enamine

2-(benzenesulfonyl)-2-chloro-1-phenylethan-1-one

  • MDL number MFCD28505996
  • Cas Number : 38009-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213891

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213891
Availability details for
Quantity
  • Availability
  • Add to Cart