ENA448968947 Enamine

1-(2-methoxy-4,6-dimethylpyridin-3-yl)methanamine dihydrochloride

  • MDL number MFCD28505997
  • Cas Number : 1803571-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213924

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213924
Availability details for
Quantity
  • Availability
  • Add to Cart