ENA448968949 Enamine

(2S)-2-amino-N,N-dimethyl-4-(methylsulfanyl)butanamide hydrochloride

  • MDL number MFCD28505999
  • Cas Number : 94903-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-144
Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213950

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213950
Availability details for
Quantity
  • Availability
  • Add to Cart