ENA448968954 Enamine

(piperidin-2-yl)methyl N-cyclopropylcarbamate hydrochloride

  • MDL number MFCD28506006
  • Cas Number : 1803588-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214066

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214066
Availability details for
Quantity
  • Availability
  • Add to Cart