ENA448968955 Enamine

(2,2-dichloroethyl)(methyl)amine hydrochloride

  • MDL number MFCD28506008
  • Cas Number : 1803594-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214089

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214089
Availability details for
Quantity
  • Availability
  • Add to Cart