ENA448968961 Enamine

1-[4-(methylamino)cyclohexyl]-2,5-dihydro-1H-pyrrole-2,5-dione; trifluoroacetic acid

  • MDL number MFCD28506019
  • Cas Number : 1803588-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214177

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214177
Availability details for
Quantity
  • Availability
  • Add to Cart