ENA448968963 Enamine

4H,5H,6H,7H-[1,2]oxazolo[4,5-c]pyridine-3-carboxylic acid hydrochloride

  • MDL number MFCD28506022
  • Cas Number : 1803587-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214193

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214193
Availability details for
Quantity
  • Availability
  • Add to Cart