ENA448968978 Enamine

[(2-amino-1-cyclopentylethyl)sulfamoyl]dimethylamine hydrochloride

  • MDL number MFCD28506042
  • Cas Number : 1803583-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214559
Availability details for
Quantity
  • Availability
  • Add to Cart