ENA448968979 Enamine

2-amino-2,3-dimethylbutan-1-ol hydrochloride

  • MDL number MFCD28506045
  • Cas Number : 149861-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199-200
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214565

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214565
Availability details for
Quantity
  • Availability
  • Add to Cart