ENA448968996 Enamine

4-(chloromethyl)-1,4-dimethylpiperidine hydrochloride

  • MDL number MFCD28506064
  • Cas Number : 1803583-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214695

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214695
Availability details for
Quantity
  • Availability
  • Add to Cart