ENA448968997 Enamine

2-(1,3-benzoxazol-2-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD28506066
  • Cas Number : 1803606-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214720

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214720
Availability details for
Quantity
  • Availability
  • Add to Cart