ENA448968998 Enamine

methyl 3-(2-aminopropan-2-yl)-1,2-oxazole-4-carboxylate hydrochloride

  • MDL number MFCD28506068
  • Cas Number : 1803608-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214814

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214814
Availability details for
Quantity
  • Availability
  • Add to Cart