ENA448969000 Enamine

4-amino-1-methyl-3-(2-methylpropyl)-1H-pyrazole-5-carboxamide

  • MDL number MFCD28506072
  • Cas Number : 268204-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214843

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214843
Availability details for
Quantity
  • Availability
  • Add to Cart