ENA448969007 Enamine

3-{[(5-chloro-2-fluorophenyl)methyl]amino}propane-1,2-diol hydrochloride

  • MDL number MFCD28506081
  • Cas Number : 1803606-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214901
Availability details for
Quantity
  • Availability
  • Add to Cart