ENA448969010 Enamine

4-chloro-2-(dimethylamino)-1,3-thiazole-5-carbonitrile

  • MDL number MFCD28506085
  • Cas Number : 1803581-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-214967

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214967
Availability details for
Quantity
  • Availability
  • Add to Cart