ENA448969024 Enamine

2-amino-3,3-dimethylbutanenitrile hydrochloride

  • MDL number MFCD28506106
  • Cas Number : 1803596-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215228

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215228
Availability details for
Quantity
  • Availability
  • Add to Cart