ENA448969025 Enamine

(1-ethyl-3,4-dihydro-1H-2-benzopyran-1-yl)methanamine hydrochloride

  • MDL number MFCD28506107
  • Cas Number : 26481-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215229

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215229
Availability details for
Quantity
  • Availability
  • Add to Cart