ENA448969031 Enamine

1-[1-(methoxymethyl)-3,4-dihydro-1H-2-benzopyran-1-yl]methanamine hydrochloride

  • MDL number MFCD28506114
  • Cas Number : 1803603-73-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215350

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215350
Availability details for
Quantity
  • Availability
  • Add to Cart