ENA448969044 Enamine

6-hydroxy-2-methylpyridine-3-carbonitrile

  • MDL number MFCD09909816
  • Cas Number : 41877-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 295-300
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213598

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213598
Availability details for
Quantity
  • Availability
  • Add to Cart