ENA448969045 Enamine

(3S)-1-(benzenesulfonyl)pyrrolidin-3-amine hydrochloride

  • MDL number MFCD28505966
  • Cas Number : 1605250-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213599

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213599
Availability details for
Quantity
  • Availability
  • Add to Cart