ENA448969050 Enamine

[1,3'-bipyrrolidin]-3-ol dihydrochloride

  • MDL number MFCD13561584
  • Cas Number : 1220038-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213215

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213215
Availability details for
Quantity
  • Availability
  • Add to Cart