ENA448969062 Enamine

(tribromomethyl)benzene

  • MDL number MFCD00461715

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213679
Availability details for
Quantity
  • Availability
  • Add to Cart