ENA448969066 Enamine

5-[methyl(prop-2-yn-1-yl)amino]pyrazine-2-carbonitrile

  • MDL number MFCD24240100
  • Cas Number : 1556142-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213691

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213691
Availability details for
Quantity
  • Availability
  • Add to Cart