ENA448969073 Enamine

2-chloro-5-(difluoromethyl)-1,3,4-thiadiazole

  • MDL number MFCD18344812
  • Cas Number : 1339678-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213698

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213698
Availability details for
Quantity
  • Availability
  • Add to Cart