ENA448969074 Enamine

4-amino-6-(dimethylamino)-1,3,5-triazine-2-carboxylic acid

  • MDL number MFCD20647430
  • Cas Number : 1379173-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213699

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213699
Availability details for
Quantity
  • Availability
  • Add to Cart