ENA448969082 Enamine

4-chloro-2-(2,2-difluoroethoxy)aniline

  • MDL number MFCD16705219
  • Cas Number : 1247621-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213709

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213709
Availability details for
Quantity
  • Availability
  • Add to Cart