ENA448969083 Enamine

1-(4-fluoronaphthalen-1-yl)ethan-1-amine

  • MDL number MFCD18706828
  • Cas Number : 1260486-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213710

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213710
Availability details for
Quantity
  • Availability
  • Add to Cart