ENA448969086 Enamine

3-methyl-4-(2-methylphenyl)-1,2-oxazol-5-amine

  • MDL number MFCD08446440
  • Cas Number : 925007-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213713

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213713
Availability details for
Quantity
  • Availability
  • Add to Cart