ENA448969089 Enamine

3-[(5-bromopyridin-2-yl)amino]butan-1-ol

  • MDL number MFCD25980870
  • Cas Number : 1549576-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213158

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213158
Availability details for
Quantity
  • Availability
  • Add to Cart