ENA448969095 Enamine

4-tert-butyl-6-chloropyrimidine

  • MDL number MFCD03731166
  • Cas Number : 3435-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 36-39
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213725

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213725
Availability details for
Quantity
  • Availability
  • Add to Cart