ENA448969101 Enamine

2-(2,6-dichlorophenoxy)-N,N-dimethylacetamide

  • MDL number MFCD28505984
  • Cas Number : 1623649-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213731
Availability details for
Quantity
  • Availability
  • Add to Cart